The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006983.1 | NP_008914.1 | 1 - 2 | 193 | 108 |
GenBank mRNA | AB010961.1 | - | 1 - 2 | 182 | 108 |
AB010962.1 | - | 1 - 2 | 207 | 108 | |
AF056200.1 | - | 1 - 2 | 170 | 108 | |
AJ005256.1 | - | 1 - 2 | 193 | 108 | |
AK297700.1 | - | 1 - 2 | 197 | 108 | |
BC025719.1 | - | 1 - 2 | 184 | 108 |