Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H2AFB1
- Entrez Gene ID
-
474382
- Gene Name
- H2A histone family member B1
- Gene Aliases
-
H2A.Bbd
- Chromosome Location
-
Chr.X: 154885042 - 154885558 on Build GRCh38
- Gene Symbol
- H2AFB2
- Entrez Gene ID
-
474381
- Gene Name
- H2A histone family member B2
- Gene Aliases
-
H2A.Bbd
- Chromosome Location
-
Chr.X: 155380706 - 155381299 on Build GRCh38
- Gene Symbol
- H2AFB3
- Entrez Gene ID
-
83740
- Gene Name
- H2A histone family member B3
- Gene Aliases
-
H2ABBD, H2AFB
- Chromosome Location
-
Chr.X: 155459419 - 155459935 on Build GRCh38
View Full Product Details