Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLA-DRB1
Entrez Gene ID
3123
Gene Name
major histocompatibility complex, class II, DR beta 1
Gene Aliases
DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
Chromosome Location
Chr.6: 32578769 - 32589836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002124.3 NP_002115.2 1 - 2 193 95
GenBank mRNA AF017440.1 - 1 - 2 95
AF291675.1 - 1 - 2 95
AJ297586.2 - 1 - 2 95
AK289463.1 - 1 - 2 95
AK291987.1 - 1 - 2 193 95
AY961062.1 - 1 - 2 95
AY961063.1 - 1 - 2 95
AY961068.1 - 1 - 2 95
AY961072.1 - 1 - 2 99 95
AY961073.1 - 1 - 2 99 95
BC024269.1 - 1 - 2 95
BC033827.1 - 1 - 2 142 95
BC108922.1 - 1 - 2 154 95
HG974574.1 - 1 - 2 99 95
HG974596.1 - 1 - 2 99 95
HG974600.1 - 1 - 2 99 95
HM067861.1 - 1 - 2 99 95
HM067862.1 - 1 - 2 99 95
HM067863.1 - 1 - 2 99 95
KJ534866.1 - 1 - 2 99 95
L02545.1 - 1 - 2 113 95
L42143.1 - 1 - 2 95
M20430.1 - 1 - 2 142 95
M20504.1 - 1 - 2 134 95
M28583.1 - 1 - 2 138 95
M28584.1 - 1 - 2 70 95
M30179.1 - 1 - 2 99 95
M33600.1 - 1 - 2 95
X02902.1 - 1 - 2 95
X03069.1 - 1 - 2 95

Gene Symbol
HLA-DRB4
Entrez Gene ID
3126
Gene Name
major histocompatibility complex, class II, DR beta 4
Gene Aliases
DR-4, DR4, DRB4, HLA-DR4B
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA M30180.1 - 1 - 2 70 95

Gene Symbol
HLA-DRB5
Entrez Gene ID
3127
Gene Name
major histocompatibility complex, class II, DR beta 5
Gene Aliases
HLA-DRB
Chromosome Location
Chr.6: 32517343 - 32530316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002125.3 NP_002116.2 1 - 2 95
XM_011514562.2 XP_011512864.1 1 - 2 95
GenBank mRNA AK314834.1 - 1 - 2 95
BC009234.2 - 1 - 2 95
M20429.1 - 1 - 2 95
M57648.1 - 1 - 2 95

View Full Product Details