The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318502.1 | NP_001305431.1 | 9 - 10 | 2204 | 74 |
NM_019892.5 | NP_063945.2 | 9 - 10 | 2207 | 74 | |
GenBank mRNA | AF187891.1 | - | 9 - 10 | 1908 | 74 |
BC028032.1 | - | 9 - 10 | 2189 | 74 |