The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291829.1 | NP_001278758.1 | 1 - 2 | 206 | 112 |
NM_001291830.1 | NP_001278759.1 | 1 - 2 | 206 | 112 | |
NR_033807.2 | 1 - 2 | 206 | 112 | ||
GenBank mRNA | AK296205.1 | - | 1 - 2 | 173 | 112 |
BX537676.1 | - | 1 - 2 | 179 | 112 | |
BX648856.1 | - | 1 - 2 | 177 | 112 |