Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF11-AS1
Entrez Gene ID
100506765
Gene Name
IGSF11 antisense RNA 1
Gene Aliases
-
Chromosome Location
Chr.3: 118943076 - 118948241 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_046230.1 1 - 2 107 84
GenBank mRNA BI829854.1 - 1 - 2 107 84


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY045493.1 - 1 - 2 137 Chr. 3 - 118943046 - 118947088 [+] on Build GRCh38 84
GenBankmRNA BG772020.1 - 1 - 2 116 Chr. 3 - 118943073 - 118947480 [+] on Build GRCh38 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC105374060 LOC105374060 -140.6kb Anti-sense Chr. - 118508411 - 118810833 on Build GRCh38
Downstream chromosome 3 open reading frame 30 C3orf30 TSCPA 201.2kb Sense Chr. - 119146150 - 119151455 on Build GRCh38

View Full Product Details