Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKT1S1
Entrez Gene ID
84335
Gene Name
AKT1 substrate 1
Gene Aliases
Lobe, PRAS40
Chromosome Location
Chr.19: 49869033 - 49878356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098632.2 NP_001092102.1 2 - 3 750 82
NM_001098633.3 NP_001092103.1 2 - 3 538 82
NM_001278159.1 NP_001265088.1 2 - 3 763 82
NM_001278160.1 NP_001265089.1 2 - 3 574 82
NM_032375.5 NP_115751.3 2 - 3 1172 82
GenBank mRNA AK055511.1 - 2 - 3 763 82
AK092610.1 - 1 - 2 1107 82
AK123687.1 - 2 - 3 1137 82
AK226004.1 - 2 - 3 1063 82
AK316603.1 - 2 - 3 404 82
BC007416.2 - 2 - 3 1058 82
BC015562.1 - 3 - 4 342 82
BC016043.1 - 2 - 3 558 82
BC051844.2 - 2 - 3 642 82

View Full Product Details