Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SEMA3B
- Entrez Gene ID
-
7869
- Gene Name
- semaphorin 3B
- Gene Aliases
-
LUCA-1, SEMA5, SEMAA, SemA, semaV
- Chromosome Location
-
Chr.3: 50267558 - 50277171 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110697.1 |
|
2 - 3 |
309 |
58 |
GenBank mRNA |
AK092182.1 |
- |
2 - 3 |
271 |
58 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BP271999.1 |
- |
2 - 3 |
428 |
|
Chr. 3 - 50273130 - 50274530 [+] on Build GRCh38 |
58 |
GenBankmRNA |
BE615357.1 |
- |
2 - 3 |
144 |
|
Chr. 3 - 50273416 - 50275595 [+] on Build GRCh38 |
58 |
GenBankmRNA |
DA693734.1 |
- |
2 - 3 |
271 |
|
Chr. 3 - 50273157 - 50274921 [+] on Build GRCh38 |
58 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 6872 |
MIR6872 |
hsa-mir-6872 |
-238.0
|
Sense |
Chr. - 50273236 - 50273297 on Build GRCh38 |
Downstream |
leucine rich single-pass membrane protein 2 |
LSMEM2 |
C3orf45 |
3.3kb
|
Sense |
Chr. - 50277907 - 50288114 on Build GRCh38 |
View Full Product Details