Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFNG
Entrez Gene ID
3458
Gene Name
interferon gamma
Gene Aliases
IFG, IFI
Chromosome Location
Chr.12: 68154770 - 68159741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000619.2 NP_000610.2 3 - 4 493 117
GenBank mRNA AB451324.1 - 3 - 4 367 117
AB451453.1 - 3 - 4 367 117
AF506749.1 - 3 - 4 301 117
AM903379.1 - 3 - 4 380 117
AY044154.1 - 3 - 4 298 117
AY255837.1 - 3 - 4 367 117
AY255839.1 - 3 - 4 301 117
BC070256.1 - 3 - 4 495 117
EF173872.1 - 1 - 2 139 117
V00543.1 - 3 - 4 458 117
X01992.1 - 3 - 4 493 117
X13274.1 - 3 - 4 475 117
X62468.1 - 3 - 4 313 117
X62469.1 - 3 - 4 313 117
X62470.1 - 3 - 4 313 117
X62471.1 - 3 - 4 313 117
X62472.1 - 3 - 4 313 117
X62473.1 - 3 - 4 313 117
X62474.1 - 3 - 4 313 117
X87308.1 - 3 - 4 307 117

View Full Product Details