Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLA-DRB1
Entrez Gene ID
3123
Gene Name
major histocompatibility complex, class II, DR beta 1
Gene Aliases
DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
Chromosome Location
Chr.6: 32578769 - 32589836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK289463.1 - 2 - 3 434 75
DQ002917.1 - 2 - 3 371 75
M11161.1 - 2 - 3 371 75
M33600.1 - 2 - 3 441 75
X03069.1 - 2 - 3 433 75

View Full Product Details