Gene Transcripts

Gene Symbol
Aqp4
Entrez Gene ID
11829
Gene Name
aquaporin 4
Gene Aliases
WCH4
Chromosome Location
Chr. 18: 15389394 - 15410977 on Build GRCm38
UniGene ID
Mm.250786
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009700.2 NP_033830.2 3 534
GenBank mRNA AF469168.1 - 3 608
AF469169.1 - 3 790
AK039777.1 - 3 537
AK044074.1 - 3 473
AK045357.1 - 2 185
AY484964.1 - 3 540
AY484965.1 - 4 724
AY484966.1 - 4 670
AY728043.1 - 3 554
AY728044.1 - 4 738
AY728045.1 - 3 666
AY803942.1 - 4 645
AY803943.1 - 5 775
AY803944.1 - 6 959
BB628835.1 - 3 535
BC024526.1 - 3 480 4985265
CT010362.1 - 2 481
U33012.1 - 2 496
U48397.1 - 2 616
U48398.1 - 2 487
U48399.1 - 4 598
U88623.1 - 3 535

View Full Product Details