Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026017.2 | NP_080293.1 | 5 | 728 | |
GenBank mRNA | AK012063.1 | - | 5 | 709 | |
AK076150.1 | - | 5 | 740 | ||
AK164602.1 | - | 5 | 670 | ||
AK193484.1 | - | 2 | 100 | ||
BC018265.1 | - | 5 | 728 | 4486464 |