Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026405.3 | NP_080681.1 | 3 | 695 | |
GenBank mRNA | AB232627.1 | - | 3 | 576 | |
AK012701.1 | - | 3 | 707 | ||
AK078874.1 | - | 3 | 387 | ||
AK088661.1 | - | 3 | 696 | ||
AK151543.1 | - | 3 | 669 | ||
AK159102.1 | - | 3 | 722 | ||
AK159901.1 | - | 3 | 722 | ||
AK163620.1 | - | 3 | 646 | ||
AY135650.1 | - | 3 | 600 |