Gene Transcripts

Gene Symbol
Dmrta2
Entrez Gene ID
242620
Gene Name
doublesex and mab-3 related transcription factor like family A2
Gene Aliases
Dmrt5
Chromosome Location
Chr. 4: 109978006 - 109983684 on Build GRCm38
UniGene ID
Mm.32825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172296.2 NP_758500.2 3 882
GenBank mRNA AK080774.1 - 3 856
AY145837.1 - 3 854

View Full Product Details