Gene Transcripts

Gene Symbol
Krtap7-1
Entrez Gene ID
71363
Gene Name
keratin associated protein 7-1
Gene Aliases
5430433J05Rik
Chromosome Location
Chr. 16: 89507703 - 89508323 on Build GRCm38
UniGene ID
Mm.158680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027771.1 NP_082047.1 1 195
GenBank mRNA AK017387.1 - 1 196
CB320982.1 - 1 178

View Full Product Details