Product Details
- SNP ID
-
rs13482113
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:28431101 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCAAATCTAGCCAACCCATAGAG[C/T]TTAGGTCTACTTTCTAGGGACTCCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Erc2
PubMed Links
Gene Details
- Gene
- Erc2
- Gene Name
- ELKS/RAB6-interacting/CAST family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177814.4 |
|
Intron |
|
|
NP_808482.2 |
View Full Product Details