Product Details

SNP ID
rs13459192
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:62505503 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTCATCATCTTCAAAATCATATCT[G/T]GCAAAGCTGTTGGGCTCTGCGGTCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Fbxo38 PubMed Links

Gene Details

Gene
Fbxo38
Gene Name
F-box protein 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_134136.3 3548 Silent Mutation NP_598897.2
Gene
Htr4
Gene Name
5 hydroxytryptamine (serotonin) receptor 4
There are no transcripts associated with this gene.

View Full Product Details