Product Details
- SNP ID
-
rs13478467
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:112937720 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCGTTCCACTCACCCCCGCAGAAG[C/T]TTGAAGAGCATACAGCCCAGGGAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Adrbk2
PubMed Links
Gene Details
- Gene
- Adrbk2
- Gene Name
- adrenergic receptor kinase, beta 2
View Full Product Details