Product Details

SNP ID
rs4228692
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:49675167 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TATTCAGATTTTCACGAGTTTACAA[A/G]TAAATCTGAGGTCCTCTGAAAGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cnot6 PubMed Links

Gene Details

Gene
Cnot6
Gene Name
CCR4-NOT transcription complex, subunit 6
There are no transcripts associated with this gene.

Gene
Gm12191
Gene Name
ribosomal protein L30 pseudogene
There are no transcripts associated with this gene.

View Full Product Details