Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF22
Entrez Gene ID
102129024
Gene Name
zinc finger protein 22
Gene Aliases
-
Chromosome Location
Chr.9: 43650127 - 43655013 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565104.2 XP_005565161.1 1 - 2 551 131

View Full Product Details