Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCT1
Entrez Gene ID
102118711
Gene Name
MYC target 1
Gene Aliases
-
Chromosome Location
Chr.4: 58934804 - 58958484 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552160.1 XP_005552217.1 1 - 2 195 109

View Full Product Details