Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUCL1
Entrez Gene ID
102119213
Gene Name
mucin like 1
Gene Aliases
-
Chromosome Location
Chr.11: 53764072 - 53765621 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015431898.1 XP_015287384.1 1 - 2 420 84

View Full Product Details