Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFCAB9
Entrez Gene ID
102114889
Gene Name
EF-hand calcium binding domain 9
Gene Aliases
-
Chromosome Location
Chr.6: 171805503 - 171815488 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558528.2 XP_005558585.1 2 - 3 338 78

View Full Product Details