Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hpn
Entrez Gene ID
15451
Gene Name
hepsin
Gene Aliases
Hlb320
Chromosome Location
Chr.7: 31098725 - 31115326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110252.2 NP_001103722.1 11 - 12 1208 78
NM_001276269.1 NP_001263198.1 10 - 11 1085 78
NM_008281.4 NP_032307.2 10 - 11 1148 78
GenBank mRNA AF030065.1 - 10 - 11 1107 78
AK002694.1 - 11 - 12 1163 78
AK156553.1 - 10 - 11 1087 78
AY234104.1 - 9 - 10 1213 78
BC138809.1 - 11 - 12 1052 78
BC145413.1 - 11 - 12 1052 78
CB949755.1 - 3 - 4 319 78

View Full Product Details