The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166406.1 | NP_001159878.1 | 16 - 17 | 2374 | 70 |
NM_001166407.1 | NP_001159879.1 | 16 - 17 | 2372 | 70 | |
NM_009004.4 | NP_033030.1 | 16 - 17 | 2268 | 70 | |
GenBank mRNA | AK051201.1 | - | 16 - 17 | 2186 | 70 |
AK051995.1 | - | 16 - 17 | 2223 | 70 | |
AK053280.1 | - | 16 - 17 | 2187 | 70 | |
AK083412.1 | - | 16 - 17 | 2228 | 70 | |
AK083697.1 | - | 16 - 17 | 2189 | 70 | |
AK084732.1 | - | 16 - 17 | 2210 | 70 | |
AK142764.1 | - | 16 - 17 | 2216 | 70 | |
AK144844.1 | - | 16 - 17 | 2186 | 70 | |
BC060608.1 | - | 16 - 17 | 2374 | 70 | |
Y09632.1 | - | 16 - 17 | 2423 | 70 |