The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271496.1 | NP_001258425.1 | 5 - 6 | 1059 | 127 |
NM_013490.4 | NP_038518.2 | 4 - 5 | 1005 | 127 | |
NR_073190.1 | 6 - 7 | 1119 | 127 | ||
GenBank mRNA | AB011002.1 | - | 4 - 5 | 864 | 127 |
AK005057.1 | - | 6 - 7 | 1120 | 127 | |
AK166465.1 | - | 4 - 5 | 946 | 127 | |
AK216139.1 | - | 1 - 2 | 111 | 127 | |
BC004719.1 | - | 2 - 3 | 195 | 127 | |
BC060218.1 | - | 4 - 5 | 968 | 127 |