Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clcn5
Entrez Gene ID
12728
Gene Name
chloride channel 5
Gene Aliases
5430408K11Rik, ClC-5, Clc5, D930009B12Rik, DXImx42e, Sfc13, T25545
Chromosome Location
Chr.X: 7153810 - 7319359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243762.1 NP_001230691.1 9 - 10 1418 82
NM_016691.4 NP_057900.3 7 - 8 1036 82
GenBank mRNA AF134117.1 - 7 - 8 1036 82
AK052990.1 - 7 - 8 1016 82
AK142227.1 - 7 - 8 967 82
AK145763.1 - 7 - 8 1005 82
BC036347.1 - 7 - 8 1008 82

View Full Product Details