Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sphk2
Entrez Gene ID
56632
Gene Name
sphingosine kinase 2
Gene Aliases
C76851
Chromosome Location
Chr.7: 45709463 - 45718002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203280.3 NP_975009.1 2 - 3 377 133
GenBank mRNA AF245448.1 - 2 - 3 327 133
AK151990.1 - 2 - 3 277 133
BC053737.1 - 2 - 3 290 133
BY256088.1 - 2 - 3 376 133

View Full Product Details