Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sphk2
Entrez Gene ID
56632
Gene Name
sphingosine kinase 2
Gene Aliases
C76851
Chromosome Location
Chr.7: 45709463 - 45718002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172561.1 NP_001166032.1 2 - 3 775 113
NM_020011.5 NP_064395.2 2 - 3 1084 113
NM_203280.3 NP_975009.1 3 - 4 846 113
GenBank mRNA AF245448.1 - 3 - 4 796 113
AF415214.1 - 1 - 2 410 113
AK004951.1 - 2 - 3 1084 113
AK146414.1 - 2 - 3 677 113
AK151990.1 - 3 - 4 746 113
AK154321.1 - 2 - 3 512 113
BC006941.1 - 2 - 3 518 113
BC053737.1 - 3 - 4 759 113
BC092084.1 - 2 - 3 533 113

View Full Product Details