The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025494.3 | NP_079770.2 | 3 - 4 | 407 | 67 |
GenBank mRNA | AB088407.1 | - | 3 - 4 | 354 | 67 |
AK006323.1 | - | 3 - 4 | 358 | 67 | |
AK075779.1 | - | 3 - 4 | 379 | 67 | |
AK129007.1 | - | 3 - 4 | 367 | 67 | |
AK151237.1 | - | 3 - 4 | 368 | 67 | |
AK168914.1 | - | 3 - 4 | 358 | 67 | |
BC010217.1 | - | 3 - 4 | 355 | 67 | |
U13839.1 | - | 3 - 4 | 350 | 67 |