Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdk2
Entrez Gene ID
18604
Gene Name
pyruvate dehydrogenase kinase, isoenzyme 2
Gene Aliases
-
Chromosome Location
Chr.11: 95026258 - 95041371 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133667.2 NP_598428.2 6 - 7 804 78
GenBank mRNA AF267660.1 - 6 - 7 765 78
AK002647.1 - 6 - 7 748 78
BC021764.1 - 6 - 7 758 78

View Full Product Details