The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010348.3 | NP_034478.1 | 5 - 6 | 1336 | 121 |
NM_146072.4 | NP_666184.2 | 5 - 6 | 1336 | 121 | |
GenBank mRNA | AK042635.1 | - | 5 - 6 | 1337 | 121 |
AK043542.1 | - | 5 - 6 | 1316 | 121 | |
AK043737.1 | - | 5 - 6 | 1300 | 121 | |
AK044250.1 | - | 5 - 6 | 1295 | 121 | |
AK044420.1 | - | 5 - 6 | 1290 | 121 | |
AK046122.1 | - | 5 - 6 | 1343 | 121 | |
AK048634.1 | - | 5 - 6 | 1307 | 121 | |
AK133667.1 | - | 5 - 6 | 1322 | 121 | |
BC031822.1 | - | 4 - 5 | 758 | 121 | |
BC049275.1 | - | 4 - 5 | 758 | 121 | |
X66118.1 | - | 5 - 6 | 1322 | 121 |