Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3bp2
Entrez Gene ID
24055
Gene Name
SH3-domain binding protein 2
Gene Aliases
3BP2
Chromosome Location
Chr.5: 34525784 - 34563639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145858.1 NP_001139330.1 2 - 3 360 86
NM_001145859.1 NP_001139331.1 2 - 3 363 86
NM_011893.3 NP_036023.3 2 - 3 260 86
GenBank mRNA AK089455.1 - 2 - 3 191 86
AK143548.1 - 2 - 3 365 86
AK150265.1 - 2 - 3 362 86
AK155103.1 - 2 - 3 217 86
AK155898.1 - 2 - 3 260 86
AK155970.1 - 2 - 3 308 86
AK156912.1 - 2 - 3 305 86
BC010198.1 - 2 - 3 198 86
BC085497.1 - 2 - 3 251 86
L14543.1 - 2 - 3 194 86

View Full Product Details