Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ormdl2
Entrez Gene ID
66844
Gene Name
ORM1-like 2 (S. cerevisiae)
Gene Aliases
0610012C09Rik
Chromosome Location
Chr.10: 128817457 - 128821632 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC002146.1 - 1 - 2 59 69

View Full Product Details