The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013892.3 | NP_038920.2 | 1 - 2 | 225 | 82 |
GenBank mRNA | AF181560.1 | - | 1 - 2 | 225 | 82 |
AK002969.1 | - | 1 - 2 | 152 | 82 | |
BC012263.1 | - | 1 - 2 | 135 | 82 |