Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exosc1
Entrez Gene ID
66583
Gene Name
exosome component 1
Gene Aliases
2610035C18Rik, 2610104C07Rik, 2610312F07Rik, AI447561
Chromosome Location
Chr.19: 41922979 - 41933418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164561.1 NP_001158033.1 1 - 2 62 79
NM_025644.4 NP_079920.1 1 - 2 62 79
GenBank mRNA AK002930.1 - 1 - 2 62 79
AK006018.1 - 1 - 2 50 79

View Full Product Details