The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025830.3 | NP_080106.1 | 10 - 11 | 1235 | 63 |
GenBank mRNA | AK004962.2 | - | 10 - 11 | 1251 | 63 |
AK053749.1 | - | 6 - 7 | 1083 | 63 | |
AK088936.1 | - | 10 - 11 | 1214 | 63 | |
AK151548.1 | - | 10 - 11 | 1229 | 63 | |
BC039921.1 | - | 10 - 11 | 1227 | 63 | |
BC048184.1 | - | 10 - 11 | 1235 | 63 | |
JN712751.1 | - | 10 - 11 | 1268 | 63 |