The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271353.1 | NP_001258282.1 | 14 - 15 | 1701 | 98 |
NM_144855.3 | NP_659104.1 | 14 - 15 | 1704 | 98 | |
NM_178224.3 | NP_835742.1 | 14 - 15 | 1704 | 98 | |
GenBank mRNA | AK077669.1 | - | 14 - 15 | 1665 | 98 |
AK085692.1 | - | 14 - 15 | 1671 | 98 | |
BC013472.1 | - | 14 - 15 | 1630 | 98 | |
BC013480.1 | - | 14 - 15 | 1640 | 98 | |
BC022144.1 | - | 14 - 15 | 1665 | 98 | |
BC026595.1 | - | 14 - 15 | 1637 | 98 |