The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008209.4 | NP_032235.1 | 5 - 6 | 1058 | 67 |
GenBank mRNA | AF010448.1 | - | 5 - 6 | 969 | 67 |
AF010449.1 | - | 5 - 6 | 784 | 67 | |
AF010450.1 | - | 5 - 6 | 784 | 67 | |
AF010451.1 | - | 2 - 3 | 1753 | 67 | |
AF010452.1 | - | 5 - 6 | 784 | 67 | |
AF010453.1 | - | - | 784 | 67 | |
AK042119.1 | - | 5 - 6 | 899 | 67 | |
AK137171.1 | - | 5 - 6 | 899 | 67 | |
AK144040.1 | - | 5 - 6 | 1003 | 67 | |
AK162712.1 | - | 5 - 6 | 1015 | 67 | |
BC026137.1 | - | 5 - 6 | 1086 | 67 | |
U94989.1 | - | 5 - 6 | 998 | 67 |