The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022313.2 | NP_071708.2 | 3 - 4 | 511 | 82 |
GenBank mRNA | AB049389.1 | - | 3 - 4 | 487 | 82 |
AF190730.1 | - | 3 - 4 | 479 | 82 | |
AF190731.1 | - | 3 - 4 | 496 | 82 | |
AK012155.1 | - | 3 - 4 | 501 | 82 | |
BC037603.1 | - | 3 - 4 | 496 | 82 | |
BC068271.1 | - | 3 - 4 | 506 | 82 |