Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bst1
Entrez Gene ID
12182
Gene Name
bone marrow stromal cell antigen 1
Gene Aliases
114/A10, A530073F09, BP-3, Bp3, Bsta1, CD157, Ly65
Chromosome Location
Chr.5: 43818880 - 43843468 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009763.3 NP_033893.2 5 - 6 625 108
GenBank mRNA AK041059.1 - 5 - 6 604 108
AK164028.1 - 3 - 4 405 108
BC033422.1 - 5 - 6 600 108
D31788.1 - 5 - 6 604 108
L32812.1 - 5 - 6 608 108

View Full Product Details