Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cdc25c
- Entrez Gene ID
-
12532
- Gene Name
- cell division cycle 25C
- Gene Aliases
-
Cdc25
- Chromosome Location
-
Chr.18: 34732993 - 34752130 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
L16926.1 |
- |
6 - 7 |
940 |
58 |
View Full Product Details