The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081459.2 | NP_001074928.1 | 6 - 7 | 2111 | 65 |
NM_001289538.1 | NP_001276467.1 | 6 - 7 | 2111 | 65 | |
NM_001289539.1 | NP_001276468.1 | 6 - 7 | 1734 | 65 | |
NM_001289540.1 | NP_001276469.1 | 7 - 8 | 1871 | 65 | |
NM_001289541.1 | NP_001276470.1 | 6 - 7 | 1734 | 65 | |
NM_001289542.1 | NP_001276471.1 | 6 - 7 | 2111 | 65 | |
NM_011363.3 | NP_035493.2 | 6 - 7 | 2111 | 65 | |
NR_110346.1 | 6 - 7 | 1682 | 65 | ||
GenBank mRNA | AF020526.1 | - | 6 - 7 | 1805 | 65 |
AF074329.1 | - | 5 - 6 | 1456 | 65 | |
AF380422.1 | - | 6 - 7 | 1805 | 65 | |
AF421138.1 | - | 6 - 7 | 1805 | 65 | |
AF421139.1 | - | 6 - 7 | 1805 | 65 | |
AK148085.1 | - | 7 - 8 | 1838 | 65 | |
AK154655.1 | - | 6 - 7 | 2045 | 65 | |
AK168439.1 | - | 6 - 7 | 1706 | 65 | |
AK170444.1 | - | 6 - 7 | 2085 | 65 | |
AK171328.1 | - | 6 - 7 | 2119 | 65 | |
AK198191.1 | - | 1 - 2 | 135 | 65 | |
BC011422.1 | - | 6 - 7 | 1656 | 65 | |
BC051978.1 | - | 6 - 7 | 1654 | 65 |