Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsm3
Entrez Gene ID
20216
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
Sa, Sah
Chromosome Location
Chr.7: 119760883 - 119784896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016870.3 NP_058566.3 14 - 15 2016 103
NM_212441.2 NP_997606.2 13 - 14 1911 103
NM_212442.2 NP_997607.2 13 - 14 1814 103
GenBank mRNA AB022340.2 - 13 - 14 1802 103
AF068246.1 - 13 - 14 1801 103
AK077247.1 - 13 - 14 1877 103
AK143946.1 - - 1814 103
AK165516.1 - 13 - 14 1788 103
AY064696.1 - 13 - 14 1887 103
BC015248.1 - 14 - 15 1976 103

View Full Product Details