The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022881.4 | NP_075019.1 | 4 - 5 | 630 | 138 |
GenBank mRNA | AB042807.1 | - | 4 - 5 | 645 | 138 |
AF302685.1 | - | 4 - 5 | 636 | 138 | |
AK036407.1 | - | 4 - 5 | 631 | 138 | |
BC099387.1 | - | 4 - 5 | 625 | 138 | |
BC132544.1 | - | 4 - 5 | 631 | 138 | |
BC132546.1 | - | 4 - 5 | 631 | 138 |