The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011540.2 | NP_035670.2 | 1 - 2 | 149 | 84 |
GenBank mRNA | AJ223854.1 | - | 1 - 2 | 149 | 84 |
AK010167.1 | - | 1 - 2 | 137 | 84 | |
BC027631.1 | - | 1 - 2 | 121 | 84 | |
BC032992.1 | - | 1 - 2 | 121 | 84 | |
Y15845.1 | - | 1 - 2 | 139 | 84 |