Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcc6
Entrez Gene ID
27421
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 6
Gene Aliases
Abcc1b, DCC, Dyscalc1, Mrp6, dyscalc
Chromosome Location
Chr.7: 45974115 - 46030486 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018795.2 NP_061265.2 17 - 18 2250 69
GenBank mRNA AB028737.1 - 17 - 18 2256 69
BC040400.1 - 17 - 18 2258 69
BC049980.1 - 1 - 2 193 69
EF109740.1 - 17 - 18 2250 69
EF109741.1 - 17 - 18 2250 69

View Full Product Details