The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021367.2 | NP_067342.1 | 4 - 5 | 330 | 87 |
NR_033206.1 | 1 - 2 | 333 | 87 | ||
GenBank mRNA | AF232937.1 | - | 4 - 5 | 330 | 87 |
AK087266.1 | - | 1 - 2 | 334 | 87 | |
BC130230.1 | - | 4 - 5 | 334 | 87 |