Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Miip
Entrez Gene ID
28010
Gene Name
migration and invasion inhibitory protein
Gene Aliases
AA553001, AI853621, D4Wsu114e, IIP45
Chromosome Location
Chr.4: 147860778 - 147868719 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025365.2 NP_001020536.2 4 - 5 980 97
GenBank mRNA AK133976.1 - 6 - 7 1132 97
AK170224.1 - 4 - 5 773 97
BC017525.1 - 4 - 5 723 97
BC024069.1 - 4 - 5 762 97
FJ618908.1 - 4 - 5 980 97

View Full Product Details