The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145374.2 | NP_663349.2 | 11 - 12 | 2818 | 61 |
GenBank mRNA | AK030941.1 | - | 4 - 5 | 1902 | 61 |
AK034623.1 | - | 3 - 4 | 2147 | 61 | |
AK079844.1 | - | 6 - 7 | 743 | 61 | |
BC020002.1 | - | 11 - 12 | 2816 | 61 | |
BC025652.1 | - | 9 - 10 | 1886 | 61 |